Print…
Pipetas monocanal, mecânicas, de volume variável ou fixo, Thermo Scientific™ F1-ClipTip™
Fornecedor: Thermo Fisher Scientific
4641180N 4641190N 4641200N 4641210N 4641220N 4641230N 4651200N 4651210N 4651220N 4651230N 4651240N 4651250N 4651260N 4651270N 4641310N 4641320N 4651280N
613-5497EA 207 EUR
613-5497 613-5498 613-5499 613-5518 613-5519 613-5520 613-5522 613-5523 613-5524 613-5525 613-5526 613-5527 613-5528 613-5529 613-6960 613-6961 613-7448
Pipetas monocanal, mecânicas, de volume variável ou fixo, Thermo Scientific™ F1-ClipTip™
Pipetas Micropipetas
The Thermo Scientific™ F1-ClipTip™ pipette features interlocking technology to ensure secure tip attachment. The pipettes work exclusively with Thermo Scientific ClipTip pipette tips, providing a unique tip interface that locks the tip in place, and ensures a complete seal with minimal tip application and ejection forces. Tips are locked firmly in place and will not fall off.
  • Silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models
  • Improved grip on the volume adjustment knob ensures effortless and precise adjustment
  • Exclusive interlock design ensures each tip is clipped securely on the F1-ClipTip until it is released with a light touch
  • Secure volume setting prevents undesirable volume drift during operation
  • 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles
  • Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of Repetitive Strain Injury (RSI)
  • Super blow-out offers 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50 µl models and below
  • Improved small volume pipetting 0,1 to 2 µl model and improved versions 1 to 10 µl models allow optimised performance in your small volume applications
  • Wide selection of ClipTip pipette tips and Compatible sizes with F1-ClipTip pipettes: 12,5 Ext, 20, 50, 200, 300, 300 Ext and 1000 µl
Thermo Scientific F1-ClipTip pipettes are warranted to be free from defects in material and workmanship for a product lifetime (10 years). This warranty is subject to conditions as stated in the attached warranty information document and require an online registration, may vary by country.
Fornecimento: Supplied with maintenance set (incl. grease and O-rings), tip fitting tool, ClipTip tip sample, shelf hanger, warning labels, calibration certificate, warranty certificate and user's guide (English, French, German, Spanish).
GLP kits are available as well.