Print…
Anti-CD20 Mouse Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 2H7]
Fornecedor: Tonbo Biosciences
65-0209-T025 65-0209-T100
65-0209-T025EA 159 EUR
65-0209-T025 65-0209-T100
Anti-CD20 Mouse Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 2H7]
Anticorpos
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation. The 2H7 antibody is reported to be cross-reactive with several non-human species including Baboon, Chimpanzee, Cynomolgus and Rhesus. Please choose the appropriate format for each application.
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: PerCP-Cyanine5.5
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human