Print…
Affinity chromatography cartridges, Ni-NTA, HisPur™
Fornecedor: Thermo Fisher Scientific
90098 90099
PIER90098EA 174 EUR
PIER90098 PIER90099
Affinity chromatography cartridges, Ni-NTA, HisPur™
Cromatografia, Colunas
Pierce™ Ni-NTA chromatography cartridges are convenient, ready to use pre-packed devices for the immobilised metal affinity chromatography purification of polyhistidine-tagged proteins from a soluble protein extract.
  • High capacity which can bind up to 60 mg of 6×His-tagged protein per ml of resin
  • Purify proteins using native or denaturing conditions
  • Same batch of resin can be reused at least five times with no loss in performance
These cartridges contain a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins. The resin effectively purifies high levels of overexpressed His-tagged fusion proteins from bacterial lysates and performs well in batch-binding and spin-column procedures at a variety of scales.
Fornecimento: Contains an accessory pack with one female Luer-Lok® adapter, one connector fitting, one column plug and bottom caps.