2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech.
Fornecedor: Thermo Fisher Scientific
|
Atenção
|
L00875.03
L00875.06
L00875.03EA
63.2
EUR
L00875.03
L00875.06
2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech.
2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate
CAS No.: 375-02-0
Molecular Formula: CF3CF2CF3CHO.xH2O
Formula Weight: 198.04(anhy)
Refractive Index: 1.3200
Beilstein Reference: 1367365
MDL No.: MFCD00150588
Fórmula:
CF₃CF₂CF₂CHO·xH₂O Temperatura de Armazenamento: Ambiente |
Número MDL :
MFCD00150588 Número CAS: 375-02-0 EINECS: 206-783-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...