Print…

Tazobactam sodium

Fornecedor: Thermo Fisher Scientific

J66226.MC J66226.03
J66226.MCEA 69.3 EUR
J66226.MC J66226.03
Tazobactam sodium
Tazobactam sodium

CAS No.: 89785-84-2 Molecular Formula: C10H11N4NaO5S Formula Weight: 322.27 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White to off-white Melting Point: 140-147° Boiling Point: 77° Solubility: Soluble in water MDL No.: MFCD00917472

Fórmula: C₁₀H₁₁N₄NaO₅S
Ponto de ebulição: 77 °C
Ponto de Fusão: 140…147 °C
Ponto de Inflamação: >110 °C (230 °F)
Número MDL : MFCD00917472
Número CAS: 89785-84-2
Merck Index: 14,9083

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR