Geldanamycin ≥99%
Fornecedor: Thermo Fisher Scientific
Atenção
|
J63397.MB
J63397.MC
J63397.MD
J63397.MBEA
149
EUR
J63397.MB
J63397.MC
J63397.MD
Geldanamycin ≥99%
Geldanamycin
Purity: 99+%
CAS No.: 30562-34-6
Molecular Formula: C29H40N2O9
Formula Weight: 560.64
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Yellow to orange
MDL No.: MFCD00274570
Applications: Benzoquinoid antibiotic that inhibits pp60src tyrosine kinase
Fórmula:
C₂₉H₄₀N₂O₉ |
Número MDL :
MFCD00274570 Número CAS: 30562-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...