Canrenoic acid potassium salt ≥98%
Fornecedor: Thermo Fisher Scientific
|
Atenção
|
J61394.03
J61394.06
J61394.14
J61394.03EA
73.1
EUR
J61394.03
J61394.06
J61394.14
Canrenoic acid potassium salt ≥98%
Canrenoic acid potassium salt
Purity: 98+%
CAS No.: 2181-04-6
Molecular Formula: C22H30KO4
Formula Weight: 396.58
Physical Form: Powder
Appearance: Pale yellow to brown
MDL No.: MFCD05662375
Fórmula:
C₂₂H₃₀KO₄ Temperatura de Armazenamento: Ambiente |
Número MDL :
MFCD05662375 Número CAS: 2181-04-6 EINECS: 218-554-9 Merck Index: 14,1750 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...