Nalpha-Acetyl-D-arginine dihydrate 98%
Fornecedor: Thermo Fisher Scientific
A18563.06
A18563.03
A18563.06EA
107
EUR
A18563.06
A18563.03
Nalpha-Acetyl-D-arginine dihydrate 98%
Nalpha-Acetyl-D-arginine dihydrate
Purity: 99%
CAS No.: 2389-86-8
Molecular Formula: C8H16N4O3.2H2O
Formula Weight: 252.29 (216.25anhy)
Melting Point: ca 270°
MDL No.: MFCD00209627
Fórmula:
C₈H₁₆N₄O₃•₂H₂O Ponto de Fusão: ∼ 270 °C Temperatura de Armazenamento: Ambiente |
Número MDL :
MFCD00209627 Número CAS: 2389-86-8 EINECS: 219-225-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...