Print…

(3,5-Dichlorophenyl)acetic acid 95%

Fornecedor: Thermo Fisher Scientific
Atenção

Sinônimos: 2-(3,5-Dichlorophenyl)acetic acid

H63293.03 H63293.06
H63293.03EA 91.8 EUR
H63293.03 H63293.06
(3,5-Dichlorophenyl)acetic acid 95%
(3,5-Dichlorophenyl)acetic acid

Purity: 95% CAS No.: 51719-65-4 Molecular Formula: C8H6Cl2O2 Formula Weight: 205.03 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD01764671

Fórmula: C₈H₆Cl₂O₂
Número MDL : MFCD01764671
Número CAS: 51719-65-4

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR