Stainless steel (Fe:Cr:Ni:Mo; 68:17:12:3 wt%), Tela metálica 200 mesh woven from 0.05mm (0.002in) Type 316
Fornecedor: Thermo Fisher Scientific
45002.RU
45002.RR
45002.CH
45002.RUEA
387
EUR
45002.RU
45002.RR
45002.CH
Stainless steel (Fe:Cr:Ni:Mo; 68:17:12:3 wt%), Tela metálica 200 mesh woven from 0.05mm (0.002in) Type 316
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 68:17:12:3 wt%
Physical Form: Wire Cloth
MDL No.: MFCD00148499