Print…
Filtros de membrana, tipo 118
Fornecedor: Sartorius
11803-100-G 11803-13-N 11803-25-N 11803-47-N 11803-50-N 11806-100-G 11806-13-N 11806-25-N 11806-47-N 11806-50-N 11806-90-G 11807-100-G 11807-13-N 11807-25-N 11807-47-N 11807-50-N 11842-100-G 11842-25-N 11842-47-N 11842-50-N 11807-90-G 11806-37-N 11842-42-N 11842-47-G 11803-90-G
512-8513EA 552 EUR
512-8513 512-8113 512-8213 512-8313 512-8413 512-8516 512-8116 512-8216 512-8316 512-8416 512-8913 512-8518 512-8118 512-8218 512-8318 512-8418 512-8511 512-8211 512-8311 512-8411 SART11807-90-G SART11806-37-N SART11842-42-N SART11842-47-G SART11803-90-G
Filtros de membrana, tipo 118
Filtros Filtros de Membrana
PTFE hydrophobic membrane filters. For filtration of air, gases, solvents and acids. Sterilisation by autoclaving or ethylene oxide.