Print…

β-Escin, white 92 - 95%, powder

Fornecedor: MP Biomedicals
Atenção

Sinônimos: 1101.2

0215794105 0215794101
ICNA0215794105EA 261 EUR
ICNA0215794105 ICNA0215794101
β-Escin, white 92 - 95%, powder
β-Escin
Peso Molecular: 1101.2 g/mol
Número MDL : MFCD00076054
Número CAS: 11072-93-8

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR