Print…
Luvas para caixas de isoladores, EPDM, AlphaTec®
Fornecedor: Ansell
85-500/9.5 85-501/11 85-502/9.5 85-501/9.5 85-500/11 85-502/11 85-503/11 85-504/9.5 85-504/11 85-505/9.5 85-503/9.5 85-505/11
112-4342EA 1170 EUR
112-4342 112-4343 112-4467 112-4344 112-4341 112-4466 112-4468 112-4471 112-4470 112-4473 112-4469 112-4472
Luvas para caixas de isoladores, EPDM, AlphaTec®
Luvas
These gloves made of diene ethylene propylene rubber (EPDM) are resistant against hydrogen peroxide solutions and common disinfecting chemicals.
  • High-quality material, in compliance with FDA CFR21 (food contact approved)
  • Comfortable and dexterous with great tactile sensitivity
  • Resist aging from exposure to oxygen, UV rays and ozone
  • Withstand temperatures up to 130 °C for repeated sterilisation
  • Halogen-free: Suitable for disposal by incineration
  • Heavy-weight version: Increased thickness (0,6 mm) for added durability
Designed for repeated autoclave sterilisation (up to 50 times), reducing the need for glove replacements. Ideal for use in pharmaceuticals and electronics.
Certificações: EN ISO 374-1 Type B (KLP), EN ISO 374-5 VIRUS, EN 388 (2111A); Cat. III