Print…
Botas de cano médio de segurança, atacadores, Entry, Safe UK
Fornecedor: U-Power
UE10123/35 UE10123/36 UE10123/37 UE10123/38 UE10123/39 UE10123/40 UE10123/41 UE10123/42 UE10123/43 UE10123/44 UE10123/45 UE10123/46 UE10123/47 UE10123/48
114-7318EA 46.2 EUR
114-7318 114-7319 114-7320 114-7321 114-7322 114-7323 114-7324 114-7325 114-7326 114-7327 114-7328 114-7329 114-7330 114-7331
Botas de cano médio de segurança, atacadores, Entry, Safe UK
Sapatos e botas
Safety ankle boots made from water resistant leather with a PU scuff cap and a slip resistant, hard-wearing PU sole.
  • Breathable 3D lining
  • Removable, anatomically shaped insole
  • Quick release lacing system
  • Natural fit width 11 for additional comfort
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Certificações: EN 20345:2011; S3, SRC