Print…
ESD safety shoes, sandals, SK Grip, Andree Grip
Fornecedor: U-Power
UK30416/35 UK30416/36 UK30416/37 UK30416/38 UK30416/39 UK30416/40 UK30416/41 UK30416/42 UK30416/43 UK30416/44 UK30416/45 UK30416/46 UK30416/47
UPOWUK30416/35EA 90 EUR
UPOWUK30416/35 UPOWUK30416/36 UPOWUK30416/37 UPOWUK30416/38 UPOWUK30416/39 UPOWUK30416/40 UPOWUK30416/41 UPOWUK30416/42 UPOWUK30416/43 UPOWUK30416/44 UPOWUK30416/45 UPOWUK30416/46 UPOWUK30416/47
ESD safety shoes, sandals, SK Grip, Andree Grip
Sapatos e botas
ESD safety sandal made from soft nubuck leather and a lightweight, extremely flexible, slip resistant PU/PU U-Grip sole.
  • Shock absorbing, anti fungal PE insole
  • PU/PU walking sole with 90° angle blocks and mulitlevel stabalising channels to for extremely high anti-slip features; TPU anti twist insert for additional support
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non metallic, puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Certificações: EN 61340 (ESD); EN 20345:2011; S1P, SRC