Print…
HPLC columns, Viva C18 (USP L1)
Fornecedor: Restek
REST9514551EA 591 EUR
REST9514551 REST9514553-700 REST9514552 REST9514351 REST9514352 REST9514561-700 REST9514315-700 REST9514332-700 REST9514563-700 REST9514562-700 REST9514333-700 REST9514562 REST9514572-700 REST9514563 REST9514365 REST9514362 REST9514561 REST9514512-700 REST9514361 REST9514365-700 REST9514525-700 REST951451E REST9514352-700 REST9514555-700 REST951431E REST951455E REST951435E REST951435B REST9514353-700 REST9514515 REST9514513-700 REST9514315 REST9514513 REST9514555 REST9514335-700 REST9514511 REST9514573-700 REST9514512 REST9514355 REST9514311 REST9514312 REST9514331 REST9514573 REST9514332 REST9514362-700 REST9514571 REST9514515-700 REST9514572 REST9514523-700 REST951452E REST951456E REST9514355-700 REST951436E REST951436B REST9514363-700 REST9514312-700 REST9514532-700 REST9514575-700 REST9514525 REST9514522 REST9514521 REST9514565 REST951450210 REST9514313-700 REST951450212 REST951453E REST951433E REST9514533-700 REST951457E REST9514565-700 REST9514522-700 REST9514535-700 REST9514552-700 REST9514535 REST9514335 REST9514575 REST9514531 REST9514532
HPLC columns, Viva C18 (USP L1)
Cromatografia, Colunas
Viva C18 are monomeric octadecylsilane columns, highly base deactivated wide pore packing that exhibits peak shape for a wide range of compounds from acidic through slightly basic. General purpose column for analyzing large molecules and biomolecules.