Print…
VWR®, Test Sieves, 300×30 mm
Fornecedor: VWR Collection
510-2593EA 266 EUR
510-2593 510-2594 510-2595 510-2596 510-2597 510-2599 510-2600 510-2601 510-2602 510-2603 510-2604 510-2605 510-2606 510-2607 510-2608 510-2609 510-2610 510-2611 510-2612 510-2613 510-2614 510-2615 510-2616 510-2617 510-2618 510-2619 510-2620 510-2621 510-2622 510-2623 510-2624 510-2625 510-2626 510-2627 510-2628 510-2629 510-2630 510-2631 510-2632 510-2633 510-2634 510-2635 510-2636 510-2637 510-2638 510-2639 510-2640 510-2641 510-2642 510-2643 510-2644 510-2645 510-2646 510-2647 510-2648 510-3846
VWR®, Test Sieves, 300×30 mm
Peneiros Peneiros de teste
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Fornecimento: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.