Print…
VWR®, Test Sieves, 450×70 mm
Fornecedor: VWR Collection
510-1693EA 666 EUR
510-1693 510-1687 510-1700 510-1699 510-1698 510-1697 510-1696 510-1701 510-1694 510-1704 510-1692 510-1691 510-1690 510-1689 510-1688 510-1695 510-1709 510-1716 510-1715 510-1714 510-1713 510-1712 510-1702 510-1710 510-1718 510-1708 510-1707 510-1706 510-1705 510-1685 510-1703 510-1711 510-1719 510-1686 510-1759 510-1766 510-1765 510-1764 510-1763 510-1762 510-1752 510-1760 510-1769 510-1758 510-1757 510-1756 510-1755 510-1754 510-1717 510-1761 510-1775 510-1781 510-1780 510-1779 510-1778 510-1767 510-1776 510-1768 510-1774 510-1773 510-1772 510-1771 510-1770 510-1751 510-1777 510-1726 510-1733 510-1732 510-1731 510-1730 510-1729 510-1753 510-1727 510-1736 510-1725 510-1724 510-1723 510-1722 510-1721 510-1720 510-1728 510-1742 510-1750 510-1749 510-1748 510-1747 510-1746 510-1745 510-1734 510-1743 510-1735 510-1741 510-1740 510-1739 510-1738 510-1737 510-1744
VWR®, Test Sieves, 450×70 mm
Peneiros Peneiros de teste
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Fornecimento: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.