Print…
VWR®, Test Sieves, 203×50 mm, with Square Holes
Fornecedor: VWR Collection
510-2569EA 185 EUR
510-2569 510-2570 510-2571 510-2572 510-2573 510-2574 510-2575 510-2576 510-2577 510-2578 510-2579 510-2580 510-2581 510-2582 510-2583 510-2584 510-2585 510-2586 510-2587 510-2588 510-2589
VWR®, Test Sieves, 203×50 mm, with Square Holes
Peneiros Peneiros de teste
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Fornecimento: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.