Print…
VWR®, Test Sieves, 200×32 mm, with Square Holes
Fornecedor: VWR Collection
510-0679EA 185 EUR
510-0679 510-0680 510-0681 510-0682 510-0683 510-0684 510-0685 510-0686 510-0687 510-0688 510-0689 510-0690 510-0691 510-0692 510-0693 510-0694 510-0695 510-0696 510-0697 510-0698 510-0699
VWR®, Test Sieves, 200×32 mm, with Square Holes
Peneiros Peneiros de teste
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Fornecimento: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.