Print…
Glass microfibre filters without binder, grade GF/F, Whatman™
Fornecedor: Whatman products (Cytiva)
1825-021 1825-024 1825-025 1825-042 1825-047 1825-055 1825-070 1825-090 1825-110 1825-125 1825-150 1825-257 1825-293 1825-915 1825-142 1825-037 1825-015
513-5272EA 84.34 EUR
513-5272 513-5241 513-5242 513-5243 513-5244 513-5245 513-5246 513-5247 513-5248 513-5249 513-5250 516-2523 516-2524 516-2531 513-0093 WHAT1825037 512-0667
Glass microfibre filters without binder, grade GF/F, Whatman™
Filtros Filtros de Fibra de Vidro
These borosilicate glass filters are ideal for measuring landfill leachate composition with a TCLP test. They can also be used for DNA binding and purification, filtering finely precipitated proteins and filtering biochemical solutions, fluids, and nucleic acids.
  • Very fine particle retention
  • Good loading capacity and flow rate
  • Binder free
Encomendar
ESPECIFICAÇÕES
Grau GF/F
Environmentally Preferable
Peso 75 g/m²
Espessura 420 µm
Retenção de partículas 0,7 µm
Velocidade de filtração 325 s (Herzberg)
Taxa de fluxo da água 41 ml/min
Débito de ar 19 s/100 ml/6,5 cm²
Max. temperature 550 °C