Print…
VWR®, Peneiros de teste, 203×32 mm
Fornecedor: VWR Collection
510-0840EA 793 EUR
510-0840 510-0841 510-0842 510-0843 510-0844 510-0845 510-0846 510-0847 510-0848 510-0849 510-0850 510-0851 510-0852 510-0853 510-0854 510-0855 510-0856 510-0857 510-0858 510-0859 510-0860 510-0861 510-0862 510-0863 510-0864 510-0865 510-0866 510-0867 510-0868 510-0869 510-0870 510-0871 510-0872 510-0873 510-0874 510-0875 510-0876 510-0877 510-0878 510-0879 510-0880 510-0881 510-0882 510-0883 510-0884 510-0885 510-0886 510-0887 510-0888 510-0889 510-0890 510-0891 510-0892 510-0893 510-0894 510-0895 510-0896 510-0897 510-0898 510-0899 510-0900 510-0901 510-0902 510-0903 510-0904 510-0905 510-0906 510-0907 510-0908 510-0909 510-0910
VWR®, Peneiros de teste, 203×32 mm
Peneiros Peneiros de teste
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Fornecimento: Mais informação do produto