Print…
Avantor® ACE® UltraCore, HPLC/UHPLC Columns, Method Development Kits, 2,5 µm
Fornecedor: Avantor
MDKU-25-1546UEA 2030 EUR
MDKU-25-1546U MDKU-25-1503U MDKU-25-1502U MDKU-25-1046U MDKU-25-1003U MDKU-25-1002U MDKU-25-0546U MDKU-25-0503U MDKU-25-0502U
Avantor® ACE® UltraCore, HPLC/UHPLC Columns, Method Development Kits, 2,5 µm
Cromatografia, Colunas
Avantor® ACE® Method Development Kits (MDK) are designed to maximize selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness. A complete range of kits for all requirements is available, including porous, solid core, HILIC and bioanalytical 300 Å column kits. These kits group together the essential column chemistries for method development and are available in a wide range of column formats.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1,7 to 5 µm) and complementary phases
  • Analytical IDs in a wide range of column lengths
  • Ideal for method development
  • Compatible with all manufacturers' LC systems
Maximum recommended pH: 1,5 to 11,0
Max. pressure: 1000 bar for analytical columns
Fornecimento: Avantor® ACE® UltraCore Method Development Kits contain Avantor® ACE® UltraCore SuperC18 and Avantor® ACE® UltraCore SuperPhenylHexyl stainless steel columns.