Print…
Anti-CD45.2 Rat Antibody (redFluor™ 710) [clone: 104]
Fornecedor: Tonbo Biosciences
80-0454-U025 80-0454-U100
80-0454-U025EA 58.6 EUR
80-0454-U025 80-0454-U100
Anti-CD45.2 Rat Antibody (redFluor™ 710) [clone: 104]
Anticorpos
The 104 antibody reacts with mouse CD45.2, also known as Ly5.2, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors.
The 104 antibody is typically used as a leukocyte marker in Ly5.2 mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. The antibody has been demonstrated to be specific for CD45.2 and is not cross-reactive with CD45.1-bearing cells.
Type: Primary
Antigen: CD45.2
Clonality:
Clone: 104
Conjugation: redFluor™ 710
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse