Print…
Anti-Hypoxanthine Phosphoribosyltransferase 1 Mouse Monoclonal Antibody [clone: 12L7]
Fornecedor: US Biological
H9800-40A-50 H9800-40A-100
USBIH9800-40A-50EA 432 EUR
USBIH9800-40A-50 USBIH9800-40A-100
Anti-Hypoxanthine Phosphoribosyltransferase 1 Mouse Monoclonal Antibody [clone: 12L7]
Anticorpos
Recommended dilution: Western Blot: 1:500-1:5000
Type: Primary
Antigen: Hypoxanthine Phosphoribosyltransferase 1
Clonality: Monoclonal
Clone: 12L7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,