Print…
Anti-Eotaxin 2 Goat Polyclonal Antibody (Biotin)
Fornecedor: US Biological
E3351-08F-25 E3351-08F-50
USBIE3351-08F-25EA 324 EUR
USBIE3351-08F-25 USBIE3351-08F-50
Anti-Eotaxin 2 Goat Polyclonal Antibody (Biotin)
Anticorpos
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml
Type: Primary
Antigen: Eotaxin 2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human,