Print…
Provetas volumétricas de mistura, formato alto, classe A, vidro borossilicato, com certificado DAkkS, BLAUBRAND®
Fornecedor: Brand
DAKKS32438 DAKKS32448
BRNDDAKKS32438EA 129 EUR
BRNDDAKKS32438 BRNDDAKKS32448
Provetas volumétricas de mistura, formato alto, classe A, vidro borossilicato, com certificado DAkkS, BLAUBRAND®
Provetas Provetas graduadas
Borosilicate glass 3.3, class A, conformity certified.
  • With DAkkS calibration certificate
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, PP stopper (except for 612-5028 and 612-5029 octagonal PE stopper)
  • Ring marks at major graduations, marks and inscriptions in high contrast blue enamel
Certificações: DIN EN ISO 4788