Print…

Anti-NQO1 Mouse Monoclonal Antibody [clone: A180]

Fornecedor: Novus Biologicals
NB200-209 NB200-209SS
NOVUNB200-209EA 635 EUR
NOVUNB200-209 NOVUNB200-209SS
Anti-NQO1 Mouse Monoclonal Antibody [clone: A180]
Anticorpos
Type: Primary
Antigen: NQO1 (NAD(P)H dehydrogenase, quinone 1)
Clonality: monoclonal
Clone: A180
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR