Print…

Anti-IgG Goat Polyclonal Antibody (CF™ 647)

Fornecedor: Biotium
20044 20044-1 20044-1MG
20044.EA 359 EUR
20044. 20044-1 20044-1MG
Anti-IgG Goat Polyclonal Antibody (CF™ 647)
Anticorpos
This is a highly cross-adsorbed goat anti-chicken IgY (H+L) secondary antibody labeled with our bright and photostable CF® dyes. To minimize cross-reactivity, the antibody has been adsorbed against bovine, goat, guinea pig, Syrian hamster, horse, human, mouse, rabbit, rat, and sheep serum.

  • Highly cross-adsorbed for specific staining with minimal background
  • Available in HRP and 10 bright and photostable CF® dyes
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

Note: Conjugates of blue fluorescent dyes like CF®405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 647
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR