Print…
Anti-cAMP Rabbit Monoclonal Antibody [clone: EP8471]
Fornecedor: Abcam
AB134901-100 AB134901-1ML
ABCAAB134901-100EA 725 EUR
ABCAAB134901-100 ABCAAB134901-1ML
Anti-cAMP Rabbit Monoclonal Antibody [clone: EP8471]
Anticorpos
Rabbit monoclonal [EP8471] to cAMP.
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: cAMP
Clonality: Monoclonal
Clone: EP8471
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar