Print…

Anti-PDGFR beta (phospho Y1021) Rabbit Monoclonal Antibody [clone: EPR2476]

Fornecedor: Abcam
AB134048-100 AB134048-1ML
ABCAAB134048-100EA 700 EUR
ABCAAB134048-100 ABCAAB134048-1ML
Anti-PDGFR beta (phospho Y1021) Rabbit Monoclonal Antibody [clone: EPR2476]
Anticorpos
Rabbit monoclonal [EPR2476] to PDGFR beta (phospho Y1021).

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PDGFR beta
Clonality: Monoclonal
Clone: EPR2476
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR