Print…

Anti-Insulin degrading enzyme/IDE Rabbit Monoclonal Antibody [clone: EPR6098(2)]

Fornecedor: Abcam
AB133561-100 AB133561-1ML
ABCAAB133561-100EA 713 EUR
ABCAAB133561-100 ABCAAB133561-1ML
Anti-Insulin degrading enzyme/IDE Rabbit Monoclonal Antibody [clone: EPR6098(2)]
Anticorpos
Rabbit monoclonal [EPR6098(2)] to Insulin degrading enzyme/IDE.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Insulin degrading enzyme/IDE
Clonality: Monoclonal
Clone: EPR6098(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR