Print…

Anti-NME2 Rabbit Monoclonal Antibody [clone: EPR8351]

Fornecedor: Abcam
AB131329-100 AB131329-1ML
ABCAAB131329-100EA 657 EUR
ABCAAB131329-100 ABCAAB131329-1ML
Anti-NME2 Rabbit Monoclonal Antibody [clone: EPR8351]
Anticorpos
Rabbit monoclonal [EPR8351] to NME2.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: NME2
Clonality: Monoclonal
Clone: EPR8351
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Human; Chinese hamster
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR