Print…

Anti-Citrate synthetase Rabbit Monoclonal Antibody [clone: EPR8066]

Fornecedor: Abcam
AB129088-100 AB129088-1ML
ABCAAB129088-100EA 650 EUR
ABCAAB129088-100 ABCAAB129088-1ML
Anti-Citrate synthetase Rabbit Monoclonal Antibody [clone: EPR8066]
Anticorpos
Rabbit monoclonal [EPR8066] to Citrate synthetase.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Citrate synthetase
Clonality: Monoclonal
Clone: EPR8066
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR