Print…
Anti-USP28 Rabbit Monoclonal Antibody [clone: EPR4250(2)]
Fornecedor: Abcam
AB110744-100 AB110744-1ML
ABCAAB110744-100EA 713 EUR
ABCAAB110744-100 ABCAAB110744-1ML
Anti-USP28 Rabbit Monoclonal Antibody [clone: EPR4250(2)]
Anticorpos
Type: Primary
Antigen: USP28 (ubiquitin specific peptidase 28)
Clonality: monoclonal
Clone: EPR4250
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar