Print…
Anti-ATP citrate lyase Rabbit Monoclonal Antibody [clone: EP704Y]
Fornecedor: Abcam
AB40793-100 AB40793-1ML
ABCAAB40793-100EA 800 EUR
ABCAAB40793-100 ABCAAB40793-1ML
Anti-ATP citrate lyase Rabbit Monoclonal Antibody [clone: EP704Y]
Anticorpos
Rabbit monoclonal [EP704Y] to ATP citrate lyase.
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ATP citrate lyase
Clonality: Monoclonal
Clone: EP704Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Human