Print…
Anti-Progesterone Receptor Rabbit Monoclonal Antibody [clone: YR85]
Fornecedor: Abcam
AB32085-100 AB32085-1ML
ABCAAB32085-100EA 800 EUR
ABCAAB32085-100 ABCAAB32085-1ML
Anti-Progesterone Receptor Rabbit Monoclonal Antibody [clone: YR85]
Anticorpos
Rabbit monoclonal [YR85] to Progesterone Receptor.
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Progesterone Receptor
Clonality: Monoclonal
Clone: YR85
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human