Print…

Anti-PKD1/PKC micro Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB99453-100 ORB99453-20
BIRBORB99453-100EA 0 EUR
BIRBORB99453-100 BIRBORB99453-20
Anti-PKD1/PKC micro Rabbit Polyclonal Antibody
Anticorpos
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: PKD1/PKC micro
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR