Print…

Anti-BIK Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB99386-100 ORB99386-20
BIRBORB99386-100EA 0 EUR
BIRBORB99386-100 BIRBORB99386-20
Anti-BIK Rabbit Polyclonal Antibody
Anticorpos
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: BIK (BCL2-interacting killer (apoptosis-inducing))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR