Print…

Anti-SDHA Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB136291-100 ORB136291-200 ORB136291-50
BIRBORB136291-100EA 0 EUR
BIRBORB136291-100 BIRBORB136291-200 BIRBORB136291-50
Anti-SDHA Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: SDHA (succinate dehydrogenase complex, subunit A, flavoprotein (Fp))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR