Print…

Anti-MAPK8 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB129654-100 ORB129654-200
BIRBORB129654-100EA 0 EUR
BIRBORB129654-100 BIRBORB129654-200
Anti-MAPK8 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: MAPK8 (mitogen-activated protein kinase 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR