Print…

Anti-FOS Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB128072-100 ORB128072-200
BIRBORB128072-100EA 0 EUR
BIRBORB128072-100 BIRBORB128072-200
Anti-FOS Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: FOS (FBJ murine osteosarcoma viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR