Print…

Anti-p15 INK Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB127851-100 ORB127851-200
BIRBORB127851-100EA 0 EUR
BIRBORB127851-100 BIRBORB127851-200
Anti-p15 INK Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: p15 INK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR