Print…

Anti-ARFGEF2 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB127699-100 ORB127699-200
BIRBORB127699-100EA 0 EUR
BIRBORB127699-100 BIRBORB127699-200
Anti-ARFGEF2 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: ARFGEF2 (ADP-ribosylation factor guanine nucleotide-exchange factor 2 (brefeldin A-inhibited))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR