Print…

Anti-c-Met Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB127499-100 ORB127499-200
BIRBORB127499-100EA 0 EUR
BIRBORB127499-100 BIRBORB127499-200
Anti-c-Met Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: c-Met
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR