Print…

Anti-AIRE Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB126718-100 ORB126718-200
BIRBORB126718-100EA 0 EUR
BIRBORB126718-100 BIRBORB126718-200
Anti-AIRE Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR