Print…

Anti-XRCC3 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB126490-100 ORB126490-200
BIRBORB126490-100EA 0 EUR
BIRBORB126490-100 BIRBORB126490-200
Anti-XRCC3 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: XRCC3 (X-ray repair complementing defective repair in Chinese hamster cells 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR