Print…

Anti-GTPase Activating Protein Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB106526-100 ORB106526-20
BIRBORB106526-100EA 0 EUR
BIRBORB106526-100 BIRBORB106526-20
Anti-GTPase Activating Protein Rabbit Polyclonal Antibody
Anticorpos
Western Blot: 1:500-1:2000; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: GTPase Activating Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR