Print…

Anti-MET Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB106351-100 ORB106351-20
BIRBORB106351-100EA 0 EUR
BIRBORB106351-100 BIRBORB106351-20
Anti-MET Rabbit Polyclonal Antibody
Anticorpos
Western Blot: 1:500-1:2000

Type: Primary
Antigen: MET (met proto-oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR