Print…

Anti-NPRL2 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
ORB101864-100 ORB101864-200
BIRBORB101864-100EA 0 EUR
BIRBORB101864-100 BIRBORB101864-200
Anti-NPRL2 Rabbit Polyclonal Antibody
Anticorpos
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: NPRL2 (nitrogen permease regulator-like 2 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR